United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
Representative photo only
Thermo Scientific Cimarec i Poly 15 stirrer 100 240V
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-04656-40
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17050-04
Innocal NIST Calibration for Stirrer Only
$115.00 USD/EACH/EACH
Usually ships in 14 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Stirrers
>
Multiposition Stirrers
>
Thermo Scientific Cimarec Electromagnetic Stirrers
Share
|
Thermo Scientific Cimarec i Poly 15 stirrer 100-240V
EW-04656-40
$1,225.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Memory function stores last speed settings
Electromagnetic drive means no moving parts and no maintenance
Use these Thermo Scientific
®
Cimarec stirrers for growing microorganisms, dissolving nutrient mediums and solids, and titration applications. Thermo Scientific Cimarec
®
Stirrers accommodate a variety of vessel shapes including Erlenmeyer flasks and beakers. Stirring speed remains constant even if viscosity of the medium changes. A three-digit LED indicates stirring speed on the 15-position models. The powder-coated stainless steel housing protects unit from spills and dirt. All stirrers include a 6-ft cord with a power supply.
Note:
Not for use in hazardous areas; not immersible.
Specifications
Product Type
Stirrer (Analog)
Speed (rpm) (low)
130
Speed (rpm) (high)
990
Top plate material
Stainless steel
Top plate dimensions (in.)
2.25 in x 2.25 in per position
(Overall top plate dimensions is 6.75 inW x 11.25 inD.)
Dimensions
9.8125 in W x 1.625 in H x 16.5 in D
Max stirring volume (Liters)
0.8
No. of stirring positions
15
Plug type
U.S. std
Model
50094596
CE Compliance
Yes
Brand
Thermo Scientific Cimarec
Manufacturer number
50094596
Power
100 to 240VAC 50/60Hz
Calibrations
Qty
/
Price
Availability
Innocal NIST Calibration for Stirrer Only
EW-17050-04
at $115.00 USD/EACH
Usually ships in 14 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
11-12 Catalog
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title