log in or register
Thermo Scientific Contact plate sterile Sabouraud dextrose agar with lecithin, polysorbate 80, pH 5.6, 10/PK
EW-14207-72
$42.75 USD/PACKAGE
Usually ships in 5 days.
Qty
  • Stringent quality control ensures high-quality performance as intended!
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
Choose from a variety of media formulations. Use this culture media for the isolation and differentiation of microorganisms from samples or specimens. Each raw component is pretested before being used, for quality assurance and consistency from lot batch. Intended for lab practice only; not intended for in vitro purposes. Plates have stacking rings which interlock to minimize sliding.
Product Type Prepared Media Plates
Description Irradiated. For cultivation of yeast and fungi.
Media Contact plate sterile Sabouraud dextrose agar with lecithin, polysorbate 80, pH 5.6
Qty/pk 10
Brand Remel
Manufacturer number R111805
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title