United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
< 16
of
25 >
Representative photo only
Thermo Scientific ERTCO Hydrometer 1 000 to 1 070 Specific Gravity 0 0005 Divisions Plain Form
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
SI-08292-16
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17107-00
NIST-Traceable Calibration; Hydrometer with test data, one-point (midpoint of Hydrometer's range)
$130.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Share
|
Sorry, this item is now discontinued but has a suggested replacement.
You can compare specifications to the suggested replacement SI-0829865
,
browse similar items
or
Contact us for more information.
Thermo Scientific ERTCO Hydrometer, 1.000 to 1.070 Specific Gravity, 0.0005 Divisions, Plain Form
SI-08292-16
$44.50 USD/EACH
Quantity Discounts
Order
Discount
Price
[X]
12
10%
$40.05/EACH
Discontinued
Qty
:
Save to shopping list
Specifications
Specific gravity range (Minimum)
1
Product Type
Hydrometers, Specific Gravity
Specific gravity range (Maximum)
1.07
Manufacturer number
15059803
Form
Plain
Length (mm)
325
Model
2560
Brand
Thermo Scientific Ertco
Accessories
Qty
/
Price
Availability
Hydrometer Case for 380 mm Length Hygrometer
EW-08285-97
at
$49.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Hydrometer with test data, one-point (midpoint of Hydrometer's range)
EW-17107-00
at $130.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title