Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Thermo Scientific GENESYS 10S Bio UV/Visible Spectrophotometer

Thermo Scientific Genesys  |   Item# EW-02654-00

$5,710.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


Usually ships in 11 days.
Qty

  • Versatile, accurate UV/Vis for life science applications
  • Pre-configured methods for nucleic acid and protein analysis
  • No warm up time needed—instant measurements from start up
  • USB connectivity to computers, printers, and data storage devices
  • Use Smart Screen to design a power-up menu that displays frequently-used tests
This spectrophotometer offers the flexibilty and performance a busy laboratory demands. The time-saving software allows you to measure DNA, RNA, and oligo concentrations. With the built-in Oligo calculator, determine molecular weight and theoretical Tm. Measure protein concentrations using Bradford, Lowry, BCA, and Biuret reagents. High-intensity xenon lamp and dual-beam optical geometry provide unsurpassed data quality.

USB connectivity allows easy access to USB memory devices. Data can be printed out using either a PCL printer or the optional internal printer.What's Included: Six-position cell holder, single-position cell holder, dust cover, and US power cord.
Product may contain trace amounts of mercury; cautions should be used in breakage, cleanup, shipping and/or disposal.
Product Type Spectrophotometer
Optics Dual-beam
Bandwidth 1.8 nm
Wavelength range 190 to 1100 nm
Wavelength accuracy ± 1.0 nm
Wavelength reproducibility ± 2.0 nm
Photometric range 0.3 to 125% T;
-0.1 A to 3 A
±9999 C
Photometric readout Transmittance, Absorbance, Concentration
Photometric noise <0.00030 @ 0 A;
<0.00055 @ 1.0 A;
<0.00085 @ 2.0 A
Photometric drift <0.0005 A/hour
Detector Dual silicon photodiodes
Output 1 USB Type A for USB external printer;
1 USB Type B for PC interface
Beam Type Double
Stray light <0.08% T @ 220;
340 nm (NaI, NaNO2);
<1.0% 198 to 200 nm (KCl)
Qty/ea 1
Source lamp Xenon
Lamp life 5 years
Display Graphical 320 x 240 pixel, Backlit LCD, 3.8" x 2.8"
(9.6 x 7.1 cm)
Dimensions (in.) 11 3/4 W x 9 3/4 H x 15 3/4 D
Dimensions (cm) 30 W x 25 H x 40 D
CE Compliance Yes
Brand Thermo Scientific Genesys

Accessories
Qty/Price
Availability

at $520.00 USD/EACH

In stock.

at $2,199.00 USD/EACH

Available for order
or call 1-888-358-4705 to order




Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Product Reviews