United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 8
of
13 >
EW-12135-35
AC 150 model shown
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-12135-35
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17001-13
NIST-Traceable Calibration; Temperature Baths
$250.00 USD/EACH/EACH
Usually ships in 21 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Baths and Circulators
>
Immersion Circulators
>
Thermo Scientific Heating Immersion Circulators
Share
|
Thermo Scientific Immersion Circulator, AC 150 thermostat with bridge (115V/60Hz)
EW-12135-35
$1,814.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Wide range of performance levels for up to 50-liter baths
Easy to use and energy efficient—rotate control head 90° for easier viewing
Advanced (AC) Controllers have all the same features as the SC models (SC Controllers feature an LCD with temperature and set point values displayed simultaneously in choice of °C, °F, or K at 0.1 or 0.01 resolution, fluid selection with predefined temperature limits, and a two-speed force pump for excellent temperature uniformity and external closed loop circulation. Additional features include five preset set point temperatures, application temperature limits, real-time clock with on/off timer), with a larger LCD, plus includes a powerful three-speed force/suction pump for external open and closed loop applications.
Additional features include seven languages (English, German, French, Spanish, Italian, Chinese, and Japanese).
What's Included:
8-mm and 12-mm hose barbs with clamps for external circulation, and a 6-ft (1.8-m) power cord with plug.
Specifications
Product Type
Circulators (Immersion)
Min temperature (º C)
Ambient
Max temperature (º C)
150
Dimensions (" W)
6-3/8
Dimensions (" H)
14-5/8
Dimensions (" D)
7-3/4
Temp control
PID
Temp sensor
Pt RTD
Temp stability
±0.01°C
High-temperature cutoff
Yes
Immersion depth
5-3/4" (14.7 cm)
Low-level cutoff
Yes
Pressure pump (Max head at 0 flow)
6.89 psi (475 mbar)
Wetted materials
stainless steel; polymers include: Polyvinylidenefluoride/hexafluoropropene (Viton®), ethylene propylene diene monomer (EPDM); polyphenylene sulfide (PPS); Polyetherimide (PEI); liquid crystal polymer (Vectra®)
Pump type
Force and Suction
Suction pump
Yes
Max flow rate (Liters/min)
20
Power (VAC)
115
Power (Hz)
60
Display
Digital
CE Compliance
Yes
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Temperature Baths
EW-17001-13
at $250.00 USD/EACH
Usually ships in 21 days.
Add to Cart
Technical Resources
Instruction Manual
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Chiller/Recirculator Selection Guide
Learn how to choose the chiller that best suits all of your cooling needs
Bath and Circulator Specifications Terminology
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title