Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Thermo Scientific Immersion Circulator, PC 200 thermostat with bridge (230VAC/50Hz)

EW-12135-55
$2,866.00 USD/EACH

Call 1-888-358-4705 for availability.
Qty

  • Wide range of performance levels for up to 50-liter baths
  • Easy to use and energy efficient—rotate control head 90° for easier viewing
Advanced (AC) Controllers have all the same features as the SC models, but also include a powerful three-speed force/suction pump for external loop applications. You get an additional five languages (Spanish, Italian, Chinese, Russian, and Japanese). They have an internal and external temperature control mode (remote sensor, NAMUR type). Features include a programmable application temperature alarm with user selected audible alarm and a “go-to-safe-state” or shut off option. The AC 200 models offer ramp programming with edit during ramp feature and a timer with real-time clock for time-critical applications. You can customize the control parameters for external and internal temperature control via serial communication (the PID-portions can be changed this way). There is also an option for extra communication for Profibus and CAN bus analog I/O—call for information.

Premium (PC) Controllers encompass all of the features of the SC and AC baths, and then provide greater control and flexibility. Pump speed adjusts to 10 different levels for turbulence control. Use the 10-key access keypad for fast and intuitive software navigation. User-level management allows you to lock in high and low temperature limitations for specific applications—prevents inadvertent changes and allows you to quickly recall saved settings. Control parameters are optimized for the application behavior, so that temperature control responds rapidly with zero over swing or under swing. For datalogging, all ring buffer data is stored for serial communication. Extra communication option for Profibus and CAN bus analog I/O—call for information.
What's Included:
8-mm and 12-mm hose barbs with clamps for external circulation, and a 6-ft (1.8-m) power cord with plug.
Product Type Circulators (Immersion)
Min temperature (º C) Ambient
Max temperature (º C) 200
Temp control PID
Temp sensor Pt RTD
Temp stability ±0.01°C
High-temperature cutoff Yes
Immersion depth 5-3/4" (14.7 cm)
Low-level cutoff Yes
Pressure pump (Max head at 0 flow) 6.89 psi (475 mbar)
Wetted materials stainless steel; polymers include: Polyvinylidenefluoride/hexafluoropropene (Viton®), ethylene propylene diene monomer (EPDM); polyphenylene sulfide (PPS); Polyetherimide (PEI); liquid crystal polymer (Vectra®)
Pump type Force and Suction
Suction pump Yes
Max flow rate (Liters/min) 20
Power (VAC) 230
Power (Hz) 50
Display Digital

Calibrations
Qty/Price
Availability

at $250.00 USD/EACH
Usually ships in 21 days.




Technical Resources
  • Instruction Manual
  • Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title