United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
3
Representative photo only
Thermo Scientific Locator Jr Cryobiological Inventory Systems
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-05094-60
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Cooling Equipment
>
Cryogenics
>
Cryogenic Storage Dewars
>
Thermo Scientific Locator Cryobiological Inventory Systems
Share
|
Thermo Scientific Locator Jr. Cryobiological Inventory Systems
EW-05094-60
$4,310.00 USD/EACH
Usually ships in 7 days.
Qty
:
Add to Cart
Save to shopping list
Store samples up to 225 days without replenishing LN
2
Compact size stores up to 3600 samples
Advanced vacuum insulation minimizes liquid nitrogen evaporation
Find the sample you need quickly and efficiently! All Thermo Scientific Locator
®
systems feature a double-walled, vacuum insulated vessel with a narrow-mouth neck that keeps liquid nitrogen losses to a minimum. Stainless steel racks fit snugly against the wall of the Thermo Scientific Locator
®
system to hold multiple racks. The rack handles rest in numbered slots for easy identification and retrieval. Each rack has individual storage locations for cryogenic boxes.
Use Thermo Scientific Locator
®
Jr. system (model 05094-60) to fit in tight spaces - this scaled-down version is only 26" in height! Thermo Scientific Locator
®
Jr. system holds four racks; each rack holds four 5-1/4" square cryogenic boxes. Select Locator 4 system (model 05094-40) to hold up to four racks; each rack holds nine 51⁄4" (13.3 cm) square cryogenic boxes. The Locator 8 system (model 05094-50) holds up to eight racks; each rack holds nine 3" (7.6 cm) square cryogenic boxes.
The Locator alarm alerts you to low liquid nitrogen level.
Order optional roller cart to easily transport your Locator system.
Specifications
Product Type
Cryobiological Inventory Systems
Capacity
51 L
Dimensions (" H)
26.5
Dimensions (dia)
22
Static holding time
60 days
No. of racks included
4
Evaporation
0.85 L/day
Max vials held
1600
Cryogenic boxes held
16
Brand
Locator
Model
CY50925
Manufacturer number
CY50925
Accessories
Qty
/
Price
Availability
Stainless steel cryogenic transfer hose, 4-ft long
EW-03773-40
at
$215.00 USD/EACH
In stock.
Add to Cart
Stainless steel cryogenic transfer hose, 6-ft long
EW-03773-42
at
$260.00 USD/EACH
In stock.
Add to Cart
Roller Cart
EW-05094-52
at
$336.00 USD/EACH
Usually ships in 5 days.
Add to Cart
Thermo Scientific Nalgene Polycarbonate CryoBox storage boxes for 100 (1.0 to 2.0 mL) vials, 10/pack
EW-06754-52
at
$114.00 USD/PACKAGE of 10
In stock.
Add to Cart
Technical Resources
Instruction Manual (70 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Isolating Genomic DNA from Whole Blood
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title