Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 
Home  > 
Discontinued Items >
  

Thermo Scientific Manual Melting Point Apparatus; with digital thermometer; 230V

Mfr# 1102DQ  |   Item# EW-03011-15

$1,780.00 USD/EACH

Discontinued
Qty

  • Built-in light and precision 6x lens enhance sample observation
  • Space-saving design uses only a small amount of valuable bench space
  • Digital units provide audio-visual alarm indicating oven temperature
These easy-to-use melting point appartuses eliminate the mess and hazards of conventional oil baths. Compact units don't take up a lot of bench space. Choose from manual or digital units. All units come with an 8-ft power cord (the 120 VAC versions have a three-prong plug; 230 VAC versions have a European plug).
Economical Manual Units have a temperature range from ambient to 500°>C. Aluminum casting conducts and radiates heat uniformly to the capillaries and thermometer. Choose from two options: Models 03011-00 and -05 come without a thermometer and include a heat guard; models 03011-10 and -15 include a digital thermometer, stand, 100 capillaries, bulb, and fuses. Order accessories below right.
Digital Unit has a temperature range from
Temp range ambient to 500°C
Heating range control continuously adjustable with built-in variable transformer
Heat-up time 25 to 400°C in 5 minutes;
400 to 500°C in 4 minutes
Cool-down time 500 to 400°C in 2 minutes; 400 to 300°C in 2 minutes;
300 to 200°C in 5 minutes
Power (Hz) 50
Power (watts) 200
Dimensions 4-1/2"W x 8-1/4"H x 5-1/2"D
Reproducibility ±1°C
Resolution Thermometer dependent
Model 1102DQ
Manufacturer number 1102DQ
Power (VAC) 230

Accessories
Qty/Price
Availability

at $128.00 USD/EACH

In stock.




Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers

Do you have questions about this product?

Get answers from real customers and in-house experts with AnswerBox.

1 Question | 1 Answer
Displaying question 1
  • Q:

    I have Thermo Scientific Mel - Temp melting point apparatus. Model # is 1301D, serial # 1877100552661. The manual does not have a picture of the apparatus. I have difficulty understanding where should the capillaries be inserted. I see a slot for thermometer.

    J. Amin
    Asked on 11/30/2010 by Anonymous

    1 answer

    • STAFF EXPERT

      A:

      The capillary openings are visible directly below the 1/2" round hole in the top of the instrument. They are also viewable by looking through the viewing glass.

      Answered on 12/2/2010 by Jack from Technical Support
Displaying question 1

Do you have a question about this product? 

Product Reviews