Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Thermo Scientific Multi-Blok Analog Dry Block Heater Base, 1-block, 120VAC
EW-36402-08
$436.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
  • Temperature uniformity within +/-0.5degC (+/-0.9degF)
  • Three-position switch—high, low, or off
  • Low-range between ambient and 60degC (140degF) and
  • high-range to 130degC (266degF)
These versatile heating systems uniformly heat a wide range of test tubes simultaneously. Systems offer different combination of blocks for your specific needs. A complete heater system consists of a standard heater base and heating blocks (sold separately). Each block heater base features a stepless thermostat dial marked from 0 to 10 for repeatable settings. A separate power switch with power-on light operates independently of the thermostat dial.
Metal-sheathed safety heaters ensure temperature uniformity and rapid heating. Heater bases have steel housing with baked enamel finish, and include 6-ft, three-wire power cord; 115 VAC models include a plug.
Max temperature (º F) 266
Watts 100
Power (VAC) 120
Max temperature (º C) 130
Dimensions 6.25 in W x 3.5 in H x 7.5 in D
Number of Blocks 1
Product Type Dry Block Heaters-Analog
Model 100
Brand Thermo Scientific
Manufacturer number 2050Q
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Do you have questions about this product?
    get answers from real customers and in-house experts with AnswerBox.
    1 Question | 1 Answer
    Displaying question 1
    • Q:
      NOT SURE HOW TO MAKE SMALL TEMP ADJUSTMENTS. PLEASE ALSO INDICATE WHAT THE PREFERRED TEMP MONITOR SYSTEM IS FOR THIS PRODUCT. THANKS
      Asked on 8/2/2013 by lllll from lllll
      1 answer
      • STAFF EXPERT
        A:
        Since this unit is analog control it is more difficult to make smaller temperature adjustments due to the knob. There is no special way to make small adjustments with it. The digital version of this heater (36402-00) is better suited for that and can be changed in 0.1 deg/C increments. As far as monitoring the temperature the units are designed to have good temperature stability, but if you wanted to have an "outside" set up monitor it you could use a thermocouple/RTD/thermistor meter and probe set up. If you have further technical questions please contact us at 800-323-4340 or techinfo@coleparmer.com.
        Answered on 8/2/2013 by Mike S from Technical Support
    Displaying question 1
    Do you have a question about this product? 