Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
log in or register

Thermo Scientific Nalgene, 1/4" Grid Case Bound Paper Lab Notebook; 1/Pk

Thermo Scientific Nalgene  |   Mfr# 6301-1000  |   Item# EW-06553-00

$39.75 USD/EACH

Quantity Discounts
Qty: 6 30 120+
Price: $35.78 $33.79 $31.80
    Laboratory notebooks feature numbered pages printed with either a green grid or green horizontal lines; additional pages include issuance page, instructions for keeping research records, and table of contents. Numbered pages have spaces for signatures and dates. Choose either Polypaper® or conventional paper. Uncoated Polypaper® is highly resistant to moisture, chemicals, tearing, and aging; works best with ballpoint pen or felt-tip marker.
    Product Type Notebooks
    No. of pages 100
    Description 8-1/2" x 11" with 1/4" grid, green
    Color Green cover
    Model 6301-1000
    Qty/pk 1
    Manufacturer number 6301-1000
    Brand Thermo Scientific Nalgene

    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers

    Do you have questions about this product?

    Get answers from real customers and in-house experts with AnswerBox.

    1 Question | 1 Answer
    Displaying question 1
    • Q:

      What are the covers made of? Are they resistant as well?
      Asked on 6/14/2011 by Joe from Richmond, VA

      1 answer

      • Staff Reviewer


        The cover is made of polyethylene, which are water proof.

        Answered on 6/16/2011 by Jen from Technical Support
    Displaying question 1

    Do you have a question about this product? 

    Product Reviews
    Customers Who Bought This Item Also Bought