United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
5 >
Representative photo only
Thermo Scientific Nalgene Labtop cooler for 12 12 to 13 mm dia tubes
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-44001-20
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Cooling Equipment
>
Cryogenics
>
Benchtop Coolers
>
Thermo Scientific Nalgene Benchtop Coolers
Share
|
Thermo Scientific Nalgene Labtop cooler for 12 (12 to 13 mm dia) tubes
EW-44001-20
$142.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
An excellent alternative to messy ice buckets for keeping reagents and enzymes cool
Lids feature a gridded write-on surface for sample identification
Stackable design saves valuable lab space
A convenient alternative to messy ice buckets. Body is molded from durable polycarbonate and filled with a nontoxic insulating gel; lid is made of clear polycarbonate (models 44001-10 and -25 have gel-filled white polycarbonate lid). Lids feature a gridded write-on surface for sample identification.
Models 44001-00, -10, and -25 include adapters to accommodate 0.5-mL tubes. Models 44001-20 and
-30 hold tubes up to 125-mm tall when lid is latched. All models have a stainless steel handle for a secure hold during transportation. Stackable design saves space.
Specifications
Product Type
Coolers
Tube size
12 to 13 mm dia
Tubes held
12
Temperature maintained
1°C, 5 hour
Max tube dimensions
12 to 13 mm dia
Dimensions
7.75 in L x 5.5 in W x 7.5 in H
Brand
Thermo Scientific Nalgene
Manufacturer number
5116-1300
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Isolating Genomic DNA from Whole Blood
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title