United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
EW-06070-22
Replacement Nalgene
®
polypropylene spigot
Click to enlarge
More Product Details
Specifications
Technical Resources
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-06070-22
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Laboratory Supplies & Consumables
>
Carboys
>
Racks, Spigots, Closures, and Accessories
>
Bottle Accessories
>
Thermo Scientific Nalgene Quick-Action Spigots
Share
|
Thermo Scientific Nalgene Replacement, polypropylene spigot
EW-06070-22
$38.50 USD/EACH
Quantity Discounts
Order
Discount
Price
[X]
6
10%
$34.65/EACH
30
15%
$32.72/EACH
120
20%
$30.80/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Use these spigots with NALGENE carboys. Spigots have 1-1/8"-12 UNF threads and accept 5/8" tubing. Spigot 06073-22 is made of polypropylene and includes an adapter for 1/4" or 1/2" ID tubing. Spigot 06073-23 is made of TEFZEL ETFE; does not include a tubing adapter. NALGENE.
Specifications
Product Type
Spigot
Cap material
Polypropylene
Material
Polypropylene
Qty/pk
1
Autoclavable
Yes
Model
6422-0010
Brand
Thermo Scientific Nalgene
Manufacturer number
6422-0010
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title