log in or register
Thermo Scientific Nalgene Storm Water Sampler Bottle, HDPE for Inorganic Analysis Sampling, Disposable
EW-99159-00
$53.00 USD/EACH
  • Ideal for economical quarterly first-flush, EPA-compliant storm water grab samples
  • Collects a full one-liter grab sample within the first 30 minutes of a qualifying rain event
  • Mount in a variety of outfall types—storm grates, ditches, and streams
  • Disposable sampler requires no decontamination
The NALGENE Storm Water Sampler combines convenience and affordability to easily meet EPA requirements for storm water grab samples.
To collect the one-liter grab sample of first flush, simply position the sample in its protective mounting tube prior to the rain event, and leave it in place until after the storm. The sampler automatically collects the sample into the bottle and shuts off after the bottle is full, preventing dilution with later runoff. After the storm, the sampler can be retrieved at your convenience.
Complete instructions included and also available for download below. Brochure available for download to the left.Mount disposable HDPE bottle with reusable Thermo Scientific Nalgene Sampler Mounting Kit 99159-12 at right.
Brand Thermo Scientific Nalgene
Manufacturer number 1100-1000
Model 1100-1000
Accessories
Qty/Price
Availability
at $64.00 USD/EACH
In stock.
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title