United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-06767-00
Thermo Scientific Nalgene
®
Wraparound Face Shield, Polycarbonate
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-06767-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Safety Products
>
Eyewear and Safety Glasses
>
Face Shields
>
Thermo Scientific Nalgene UV-Protective Wraparound Face Shield
Share
|
Thermo Scientific Nalgene Wraparound Face Shield, Polycarbonate
EW-06767-00
$111.00 USD/EACH
Usually ships in 7 days or sooner.
Qty
:
Add to Cart
Save to shopping list
Full-length, wrap-around shield sits away from face for easy breathing
Optically clear window of 0.06" thick polycarbonate meets ANSI standard Z87.1protects against UV radiation from 200 to 360 nm. Adjustable headband.
Compliance:
ANSI Z87.1, 29 CFR Part 1910.1030
Specifications
Product Type
Face Shields
Color
Gray
Lens
Clear
Coating
UV protection
Compliance
ANSI Z87.1; 29 CFR Part 1910.1030
Brand
Thermo Scientific Nalgene
Model
6355-0001
Manufacturer number
6355-0001
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Workplace eye safety is as simple as 1 – 2 – 3
Workplace Safety Quiz
Ten Ways to Prevent Eye Injuries at Work
Show All...
Workplace Eye Safety
Safety Eyewear Selection Guide
Eye Safety- Emergency Response and Disaster Recovery
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title