United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
4 >
EW-83008-02
Shown with the included iD1 transmission accessory.
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
More info
Related technical articles
Service & Support
Related media
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-83008-02
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Material Testing & Physical Testing
>
Spectroscopy and Spectrophotometers
>
Infrared (IR) Spectroscopy
>
Thermo Scientific Nicolet iS5 FT-IR Spectrometer
Share
|
Thermo Scientific Nicolet iS5 FT-IR Spectrometer, with KBr Windows, English
EW-83008-02
$11,275.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Ideal for laboratory analysis, raw material identification, and finished product inspection
Compact and light enough for on-site or multi-location use
Field-proven optics housed in a sturdy magnesium-alloy frame
Thermally-controlled diode laser provides years of worry-free use
User-accessible parts minimize service costs
Magnesium-alloy construction provides excellent mechanical characteristics for stiffness, thermal properties, vibrational dampening and reduced weight
Tightly sealed and desiccated optical bench protects internal components from high humidity and aggressive chemicals
Internal diagnostics with electronic humidity and heat sensors warn of excessive humidity or temperature
The Nicolet iS5 spectrometer is designed to withstand a wider range of temperature and humidity conditions than those found in a typical laboratory. Vibration, electromagnetic interference (EMI), dust, and even tilt were all considered in the design of this instrument.
The Nicolet iS5 spectrometer delivers outstanding performance in raw materials inspection and in the identification of unknowns. An intuitive user interface and high signal-to-noise ensure accurate results with no need for spectroscopy expertise. Different sensitivity levels account for allowable variations in raw materials. Normal sensitivity QCheck gives you high confidence in PASS/FAIL material inspections of natural and other small variation products; the high sensitivity QCheck accommodates high confidence PASS/FAIL material inspection of synthetic and other highly consistent products.
For compounds and mixtures, when PASS/FAIL does not provide sufficient information, the OMNIC
Specta
software includes a patented algorithm that greatly improves the contaminants analysis and identifies all compounds in a multiple-ingredient material. Additional software features include data collect, stacked/overlaid displays, cursors, peakpick, peak height/area, subtraction, electronic lab notebook, and macro programming.
Performance verification is ensured by automatic testing to ASTM E1421 specifications to meet ISO and GMP requirements. Optical alignment is also automatic, with dynamic alignment for continuous signal optimization.
The small footprint takes up minimal lab space with dimensions of 13.5"W x 10.9"D. The small size also allows for movement from location to location, weighing just 22 lb (10 kg).
The iS5 includes the iD1 transmission accessory for samples in liquid cells, gas cells, pellet holders, or cuvettes; supporting cells up to 10-cm in length. The IR source, power supply, desiccants and sample compartment windows are all easily user-replaceable without exposing internal componentsreducing maintenance and ownership costs. Order optional high-performance ATR sampling accessories below.
What's Included:
iD1 transmission accessory; for cells up to 10 cm in length.
Specifications
Product Type
Infared Spectroscopes
Sensitivity
22,000:1 (1 minute, 4 cm
-1
)
Spectrum range
7800 to 350 cm
-1
Detector
dTGS room-temperature
Output
USB 2.0
Resolution
< 0.8 cm
-1
Dimensions
13.5" x 10.2" x 10.9"
(35 x 26 x 28 cm)
Source lamp
Mid-IR Ceramic emitter
CE Compliance
Yes
Brand
Thermo Scientific Nicolet
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
More info
Product Specifications - English
Related technical articles (Roll over for summary)
Real or Phony: An analysis of artwork for authenticity
Seeing the Light: An Overview of Visible and UV-VIS Spectroscopy
Spectrophotometer Selection Guide
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Related media
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title