log in or register
Thermo Scientific Orion Thermo /COD165 reactor
EW-99581-35
$1,115.00 USD/EACH
Usually ships in 7 days.
Qty
  • Ideal for environmental and wastewater monitoring
  • Simultaneous digestion of 25 samples saves time
  • For a complete system order COD reactor (99581-35), COD test kits (99581-40,-42, or -44), and COD colorimeter (99581-20)
Works with Thermo Scientific Orion COD kits or any 16 mm COD vial. The simple two-button interface, combined with the auto-shutoff feature, easily sets the reactor to digest up to 25 vials at once. Preset reaction times of 30, 60, or 120 minutes eliminate the need for an external timer. Use continuous mode for other digestion times. Set the temperature to 100°C, 120°C, 150°C, 160°C, or 165°C to start the reactor.
Accessories
Qty/Price
Availability
at $25.60 USD/PACKAGE of 4
In stock.
at $38.00 USD/EACH
Usually ships in 11 days.
at $38.00 USD/EACH
Usually ships in 12 days.
at $38.00 USD/EACH
Call 1-888-358-4705 for availability.
at $45.00 USD/EACH
In stock.
at $50.00 USD/EACH
In stock.
Technical Resources
  • Instruction Manual
  • Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title