Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Thermo Scientific Orion Versa Star with Two pH/ISE Modules

Thermo Scientific Orion  |   Item# WU-58825-89

$2,261.00 USD/EACH
$2,147.95 USD/EACH

Are you a reseller or buying in bulk? Call us today.


Usually ships in 9 days.
Qty

  • Modularity allows you to configure the meter how you need it--for now and for later
  • Customize the four measurement channels with five different parameter modules
  • Large color graphic LCD with intuitive multilanguage interface
  • Non-volatle memory holds up to 2000 data points with time and date stamp
  • Easy-to-operate USB and RS232 ports and data transfer software included

  • Up to 5 point calibration with automatic recognition for USA/NIST and DIN buffers
  • Editing calibration errors with a one point recalibration
  • Advanced ISE features such as auto-blank, low-concentration stability range, and incremental techniques
The VERSA STAR advanced meter provides the versatility to operate from one to four measurement channels at the same time. The five parameter modules available for the channels including; pH, pH/ISE, Conductivity, DO/RDO, and pH with LogR. Customize the meter you need with the options of meter only and modules, a meter with module package, or a full kit with meter, modules, electrode and solutions.

This versatile meter has a large color graphic LCD display. The informative display shows readings with temperature, connections, time, date, sample ID, user ID and calibration data. Plain language screen prompts, soft keys, and a multilanguage interface supports English, Spanish, French, Italian, German, and Chinese.

Non-volatle memory holds up to 2000 data sets with time and date stamp. Easy-to-operate USB and RS232 ports and data transfer software take stress out of data management.
Add two pH/ISE/Temperature modules. Order probes separately.
What's Included:
VERSA STAR meters include: an electrode arm with redesigned holder, and a universal power adapter.
Product Type Multiparameter Meter
Meter style Benchtop
Range (pH) -2.000 to 20.000
Range (mV) ± 2000
Range (ISE) 0 to 19999 ppm
Range (Temperature °C) -5 to 105
Resolution (pH) 0.1/0.01/0.001
Resolution (Temperature) 0.1
Resolution (mV) 0.1/1
Resolution (ISE) Up to 3 digits
Accuracy (pH) ±0.002 pH
Accuracy (Temperature) ±0.1°C
Accuracy (mV) ±0.2 mV or 0.05%, whichever is greater
Accuracy (ISE) ±0.05% of reading
Temperature compensation Automatic or manual
Display LCD, graphic
Interface USB or RS232
Calibration Up to 6 points
Waterproof Yes
Qty/ea 1
Buffer recognition USA, NIST, DIN
Power AC adapter or four AA batteries(optional)
Datalogging (points) 2000
CE Compliance Yes
Brand Thermo Scientific Orion
Dimensions (" W) 7
Dimensions (" H) 9.75
Dimensions (" D) 3.375
Model VERSA STAR W 2 pH/ISE Modules

Accessories
Qty/Price
Availability

at $572.00 USD/EACH

In stock.

at $9.25 $7.40 USD/PACKAGE of 4

In stock.

at $110.00 USD/EACH

In stock.

at $607.00 $576.65 USD/EACH

Usually ships in 9 days.

at $1,003.00 $952.85 USD/EACH

Usually ships in 9 days.

at $992.00 $942.40 USD/EACH

Usually ships in 9 days.

at $1,134.00 $1,077.30 USD/EACH

Usually ships in 9 days.

at $979.00 $930.05 USD/EACH

Usually ships in 9 days.

at $178.00 USD/EACH

In stock.

at $60.50 USD/EACH

In stock.
Related products
Qty/Price
Availability

at $17.75 USD/PACKAGE
In stock.
Calibrations
Qty/Price
Availability

at $126.00 USD/EACH
Usually ships in 6 days.




Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers

Do you have questions about this product?

Get answers from real customers and in-house experts with AnswerBox.

1 Question | 1 Answer
Displaying question 1
  • Q:

    I have a few questions about the datalogging. Can the meter be hooked up to a computer and have the meter log data continuously? Alternatively, can the time interval for the logging be set manually?
    Asked on 5/24/2012 by Anonymous

    1 answer

    • STAFF EXPERT

      A:

      Software is not yet available for this unit, but will be introduced within 30 business days. Hyper Terminal is available free of charge through Microsoft right now to export data through the RS232 cable. The data is not transmitted continuously. It send data about every 5 seconds because the unit resets itself in those intervals. The time interval for logging can be set manually with 5 seconds as the shortest interval.

      Answered on 5/25/2012 by Rita from Customer Service
Displaying question 1

Do you have a question about this product? 

Product Reviews