dummy
EW-58825-95
Usually ships in 6 days.
Qty:
- Modularity allows you to configue the meter how you need it--for now and for later
- Customize the four measurement channels with five different parameter modules
- Large color graphic LCD with intuitive multilanguage interface
- Non-volatle memory holds up to 2000 data points with time and date stamp
- Easy-to-operate USB and RS232 ports and data transfer software included
- Up to 5 point calibration with automatic recognition for USA/NIST and DIN buffers
- Editing calibration errors with a one point recalibration
- Selectable cell constant for 2- or 4-cell conductivity probes
- Selectable reading reference temperature of 5, 10, 15, 20 or 25°C
- Choice of practical salinity or natural sea water curves for salinity
- Accepts both polarographic and RDO(optical/luminessence) probes
- Automatic temperature and barometic pressure compensation, manual salinity compensation
The VERSA STAR advanced meter provides the versatility to operate from one to four measurement channels at the same time. The five parameter modules available for the channels including; pH, pH/ISE, Conductivity, DO/RDO, and pH with LogR. Customize the meter you need with the options of meter only and modules, a meter with module package, or a full kit with meter, modules, electrode and solutions.
This versatile meter has a large color graphic LCD display. The informative display shows readings with temperature, connections, time, date, sample ID, user ID and calibration data. Plain language screen prompts, soft keys, and a multilanguage interface supports English, Spanish, French, Italian, German, and Chinese.
Non-volatle memory holds up to 2000 data sets with time and date stamp. Easy-to-operate USB and RS232 ports and data transfer software take stress out of data management.
Adds one pH/ISE, one Conductivity, and one DO/RDO measurement modules. Probes sold separately.
What's Included:
VERSA STAR meters include: an electrode arm with redesigned holder, and a universal power adapter.
| Product Type |
Multiparameter Meter |
| Meter style |
Benchtop |
| Dimensions (" W) |
7 |
| Dimensions (" H) |
9.75 |
| Dimensions (" D) |
3.375 |
| Range (pH) |
2.000 to 20.000 |
| Range (mV) |
± 2000 |
| Range (Conductivity) |
0.001µ S to 3000 mS |
| Range (TDS) |
0 to 200 ppt |
| Range (Salinity) |
0 to 80 ppt |
| Range (Resistivity) |
2 ohm to 100 mega-ohm |
| Range (ISE) |
0 to 19999 ppm |
| Range (Dissolved oxygen) |
0 to 90 mg/L; 0 to 600 % |
| Range (Temperature °C) |
5 to 105 |
| Resolution (pH) |
0.1/0.01/0.001 |
| Resolution (Temperature) |
0.1 |
| Resolution (mV) |
0.1/1 |
| Resolution (Conductivity) |
0.001, 0.01, 0.1 µS; 0.01, 0.1 mS |
| Resolution (TDS) |
Up to 4 digits |
| Resolution (Resistivity) |
Up to 4 digits |
| Resolution (ISE) |
Up to 3 digits |
| Resolution (Salinity) |
Up to 4 digits |
| Accuracy (pH) |
±0.002 pH |
| Accuracy (Temperature) |
±0.1°C |
| Accuracy (mV) |
±0.2 mV or 0.05%, whichever is greater |
| Accuracy (Resistivity) |
±0.5% ±1 digit |
| Accuracy (ISE) |
±0.05% of reading |
| Accuracy (Conductivity) |
±0.5% full-scale ±1 LSD |
| Accuracy (TDS) |
± 0.5% full-scale |
| Accuracy (Salinity) |
± 0.1 ppt |
| Accuracy (Dissolved Oxygen) |
±0.2 mg/L |
| Temperature compensation |
Automatic or manual |
| Display |
LCD, graphic |
| Interface |
USB or RS232 |
| Calibration |
Up to 6 points |
| Waterproof |
Yes |
| Buffer recognition |
USA, NIST, DIN |
| Power |
AC adapter or four AA batteries(optional) |
| Datalogging (points) |
2000 |
| CE Compliance |
Yes |
| Brand |
Thermo Scientific Orion |
| Model |
VERSA STAR W Multi Modules |
at $528.00 USD/EACH
In stock.
at $7.50 USD/PACKAGE of 4
In stock.
at $395.00 USD/EACH
Usually ships in 11 days.
at $496.00 USD/EACH
In stock.
at $729.00 USD/EACH
Call 1-888-358-4705 for availability.
at $746.00 USD/EACH
Usually ships in 14 days or sooner.
at $94.00 USD/EACH
In stock.
at $536.00 USD/EACH
Usually ships in 6 days.
at $867.00 USD/EACH
Usually ships in 6 days.
at $867.00 USD/EACH
Usually ships in 6 days.
at $1,015.00 USD/EACH
Usually ships in 6 days.
at $913.00 USD/EACH
Usually ships in 6 days.
at $152.00 USD/EACH
Usually ships in 6 days.
at $51.00 USD/EACH
Usually ships in 6 days.
at $115.00 USD/EACH
Usually ships in 9 days.
at $115.00 USD/EACH
Usually ships in 9 days.
Technical Resources
Related technical articles (Roll over for summary)
Show All...
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334