dummy
EW-05723-02
- Orion-designed for optimum performance and the most comprehensive ISE selection
- EPA approved
ionplus® Sure-Flow® Combination Ion-Selective Electrodes are easier to use than reference and half-cells—eliminating fumbling and tangles from multiple cables. Sure-flow® technology provides stable, reproducible results. The easy-to-clean design reduces the risk of inaccurate readings due to clogging of the reference junction.
All electrodes include a BNC connector, a 3-ft cable, and manual. Complete systems require-the purchase of a standardizing solution, filling solution for the reference electrode, ionic strength adjustor, and pH adjusters.
Chloride, cyanide, Sulfide, nitrate, and fluoride are approved for EPA methods for analysis of wastewater and for NPDES permitting.Electode type: Solid state
Concentration range: 10-8 M to saturated; 0.02 ppm to saturated
Temperature/ pH range: 0 to 80°C/ 5 to 11
Interferences: OH
| Product Type |
Solid-State |
| pH range |
5 to 11 |
| Temperature range |
0 to 80°C |
| Connector |
BNC (Waterproof when used with Star Series meters) |
| Model |
9609BNWP |
| Brand |
Ionplus |
at $79.00 USD/EACH
Usually ships in 7 days or sooner.
at $69.00 USD/EACH
Usually ships in 10 days or sooner.
at $132.00 USD/GALLON
In stock.
at $140.00 USD/EACH
In stock.
at $89.00 USD/CARTON of 5
In stock.
Technical Resources
Related technical articles (Roll over for summary)
Show All...
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334