Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Thermo Scientific PRECISION Four-hole steaming bath, 240V

EW-24100-02
$1,461.00 USD/EACH

Call 1-888-358-4705 for availability.
Qty

  • Concentric rings can be removed from inner cover outward to accommodate various vessel shapes and sizes
  • Copper clad immersion heaters provide bath temperatures up to 100°C
  • Easy to operate with a 4-position switch and 3 heat settings
Thermo Scientific PRECISION Concentric Ring Steaming Baths are designed to gently warm samples, evaporate liquids, and melt solids such as agar. Concentric rings can be removed from inner cover outward to accommodate various vessel shapes and sizes. Four- and eight-hole models enlarge in 1” increments from 7/8” at the center, to a maximum opening diameter of 5 and 6 inches, respectively. Copper clad immersion heaters provide bath temperatures up to 100°C, with three variable heat settings.

Maintain bath depth at desired level with adjustable water level regulator. Epoxy polyester-coated steel support frame is resistant to corrosion. Stainless steel flanged cabinet top can be installed in the benchtop providing a permanent station. Baths must be hard- wired to the electrical system by a qualified electrician.
What's Included:
constant water level device, adjustable drain tube, and support frame.
Product Type Water Bath
Bath type Special Purpose Water Baths
Capacity (Liters) 11.3
Heater settings: Low 275W
Heater settings: Medium 550W
Heater settings: High 1100W
Power (VAC) 240
CE Compliance Yes

Calibrations
Qty/Price
Availability

at $250.00 USD/EACH
Usually ships in 21 days.




Technical Resources
  • Instruction Manual.pdf
  • Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title