Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Sorry, this item is now discontinued.browse similar itemsor Contact us for more information.
Thermo Scientific SPECTRONIC standards, set of 10
Thermo Scientific  |   Mfr# 333150-000  |   Item# EW-02650-57
$972.00 USD/SET of 1
Discontinued
Qty
  • Easily and accurately perform certified spectrophotometer evaluations in your own laboratory
  • Designed for use in visible light ranges
  • ISO17025 accredited for testing and calibration
  • Easily and accurately perform certifified spectrophotometer evaluations in your own laboratory
  • Standards are designed for use in visible light ranges
  • Standards are ISO17025 accredited for testing and calibration
Check the performance and accuracy of your spectrophotometer right in your own lab. Simply insert standards into your spectrophotometer's cell holder. Standards include:
0% transmittance: the opaque 0%T standard is used to test for light leaks in your spectrophotometer.
Wavelength 590 nm: the wavelength accuracy standard has a transmittance peak near 515 nm which is certified as a wavelength accuracy standard and is NIST traceable. The symmetry of the peak means that the tests of wavelength accuracy are independent of the spectral bandwidth (up to 20 nm) of the instrument being tested. In addition, the testing peak of 515 nm is located at that point when the tungsten lamp shows the flattest energy measurement in response to wavelength change.
Stray radiant energy: each of these three standards is essentially opaque at its test wavelength and highly transmitting at longer wavelengths. The test wavelength is chosen (220 nm, 340 nm, and 400 nm) to correspond to points of low energy for both tungsten and deuterium lamps, readily revealing any stray energy for both the visible and UV regions.
Photometric performance: these 10% T and 50% T neutral density filters test spectrophotometric accuracy, precision, and linearity. No spectral bandwidth is required. The control standard verifies that the holder itself does not affect the 100%T reading.
Optical alignment: optical alignment in the sample compartment can be evaluated by using the control standard and photosensitive paper. The paper records the position of the light beam in the cuvette holder.
Check the performance and accuracy of your spectrophotometer right in your own lab. Simply insert standards into your spectrophotometer's cell holder. Standards include:
0% transmittance: the opaque 0%T standard is used to test for light leaks in your spectrophotometer.
Wavelength 590nm: the wavelength accuracy standard has a transmittance peak near 515 nm which is certified as a wavelength accuracy standard and is NIST traceable. The symmetry of the peak means that the tests of wavelength accuracy are independent of the spectral bandwidth (up to 20 nm) of the instrument being tested. In addition, the testing peak of 515 nm is located at that point when the tungsten lamp shows the flattest energy measurement in response to wavelength change.
Stray radiant energy: each of these three standards is essentially opaque at its test wavelength and highly transmitting at longer wavelengths. The test wavelength is chosen (220 nm, 340 nm, and 400 nm) to correspond to points of low energy for both tungsten and deuterium lamps, readily revealing any stray energy for both the visible and UV regions.
Photometric performance: these 10% T and 50% T neutral density filters test spectrophotometric accuracy, precision, and linearity. No spectral bandwidth is required. The control standard verifies that the holder itself does not affect the 100%T reading.
Optical alignment: optical alignment in the sample compartment can be evaluated by using the control standard and photosensitive paper. The paper records the position of the light beam in the cuvette holder.
Product Type Spectroscopy Calibration Standards
Applications Visible Light Spectroscopy
Model 333150-000
Manufacturer number 333150-000
Brand Thermo Scientific
Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Do you have questions about this product?
    Get answers from real customers and in-house experts with AnswerBox.
    6 Questions | 7 Answers
    Displaying questions 1-6
    • Q:
      why there are two sets of 10% T and 50% T neutral density filters for photometric performance
      Asked on 12/19/2012 by Amb from Rwland hts, CA
      1 answer
      • STAFF EXPERT
        A:
        These 10%T and 50%T Neutral Density filters test spectrophotometric accuracy, precision and linearity. To test linearity there has to be more than one test point. Since the transmittance of this glass is extremely constant with changes in wavelength near 590 nm, spectrophotometric errors can be distinguished from wavelength errors. No spectral bandwidth compensation is required. The Control Standard verifies that the holder itself does not affect the 100%T reading.
        Answered on 12/21/2012 by Jen from Technical Support
    • Q:
      I am interested in getting these recertified. How can I do this.
      Asked on 12/29/2010 by Anonymous from Winston-Salem, NC
      1 answer
      • Staff Reviewer
        A:
        Recertification may be completed through the return and purchase of part number 00256IO.
        Answered on 1/21/2011 by Jen from Technical Support
    • Q:
      Does this come with a instruction manual? If so, how can I get one, since we already have the set?
      Asked on 11/18/2010 by Jan from Medicine Hat, AB
      1 answer
      • STAFF EXPERT
        A:
        No, this product does not come with an instructional manual. There should be instructions for calibration in the manual for the meter.
        Answered on 11/19/2010 by Jack from Technical Support
    • Q:
      Can I purchase just one or a few standards, instead of the set?
      Asked on 1/12/2010 by cmbogda from North Branch, NJ
      1 answer
      • STAFF EXPERT
        A:
        Standards include: 0% Transmittance, Wavelength 590nm, Stray Radiant Energy, Photometric Performance and Optical Alignment.
        Is there a specific standard you are looking for? Please contact our technical application specialists for additional assistance (contact information can be found on the product web page).
        Answered on 1/14/2010 by Jen from Technical Support
    • Q:
      What standards are included?
      Asked on 8/29/2009 by EL QUIMICO from IRAPUATO,GUANAJUATO.MEXICO
      1 answer
      • STAFF EXPERT
        A:
        Standards include: 0% Transmittance, Wavelength 590nm, Stray Radiant Energy, Photometric Performance and Optical Alignment.
        Answered on 8/31/2009 by Zeke from Technical Support
    • Q:
      Can I use this for our 20-D Milton Roy Spectrophotometer
      Asked on 8/10/2009 by Nette from Philippines
      2 answers
      • A:
        Before purchasing ANY kit please check that tolerances of all measurements provided in the kit's certificate (so-called Expanded Uncertainties) are not too loose for adequate testing of performance of your spectrophotometer. Another words, higher quality spectrophotometer you have, better tolerances are required for calibration standards to test the spectrophotmeter adequately. The best calibration standards and kits are offered by NIST and by only few other companies in the world which manufacture NIST compliant and/or NIST compatible calibration standards and kits. ("NIST traceability" statement is used by many manufacturers of kits and standards but it is NOT the same as "NIST compliance"/"NIST compatibility"). Certainly, these standards are much more expensive comparably to the cost of this kit.
        Answered on 2/18/2014 by Sci from NY
      • STAFF EXPERT
        A:
        These standards should work with most spectrophotometers, not just Thermo brand products.
        Answered on 8/11/2009 by Jen from Techncial Support
    Displaying questions 1-6
    Do you have a question about this product? 
    Product Reviews