dummy
EW-02905-05
- High Performance Dialysis require just half the time of dialysis tubing for sample size from 100µl to 30mL
- Reduces preparation time
- The innovative cassette design does not require the use of knots or clips that can lead to leaking sample loss.
- Color coded translucent frame permits smooth sample withdrawal of sub-filter volumes ¨C removing every last drop is easy.
- High surface area/sample volume ratio will dialyze twice as fast as dialysis via conventional tubing.
- Available in a variety of sizes
Slide-A-Lyzer® dialysis cassette is ideal for low molecular weight contaminant removal, buffer exchange, desalting and concentration. The cassette membrane is composed of low-binding regenerated cellulose and features a hermetically sealed sample chamber to maintain the highest possible sample retention. Sample introduction and removal are easily accomplished by penetrating the gasket with a hypodermic needle attached to a syringe. The gasket imparts resealability to the needle puncture, ensuring that no sample is lost from the cassette during dialysis.Ready-to-use dialysis devices offer conveniences with preparing, loading and retrieving samples. These devices also increase recovery and decrease sample loss, while increasing sampling efficiency. Choose the ready-to-use device that fits your application. The Micro Float-A-Lyzer is ideal for small sample volumes, below 500µL. The Tube-A-Lyzer uses a flow-through buffer chamber to increase transfer rate and drastically reduce run times. The Float-A-Lyzer G2 makes sample loading and retrieval easy, and includes color coded cap colors for different Molecular Weight Cut-Offs (MWCO). The Thermo Scientific Slide-A-Lyzer has a high surface area to sample volume ratio to increase run times.
| Product Type |
Ready-to-use Dialysis Device |
| Material |
Regenerated Cellulose |
| Membrane MWCO (Daltons) |
3500 |
| Min sample volume (mL) |
3 |
| Max sample volume (mL) |
12 |
| Color |
Pink |
| Qty/pk |
8 |
| Brand |
Thermo Scientific |
| Manufacturer number |
66110 |
| Model |
Slide-A-Lyzer Dialysis Cassettes |
at $34.00 USD/EACH
Usually ships in 4 days.
at $47.00 USD/EACH
Usually ships in 15 days.
at $27.00 USD/EACH
Usually ships in 15 days.
at $27.00 USD/EACH
Usually ships in 5 days.
at $28.00 USD/EACH
Usually ships in 5 days.
at $29.00 USD/EACH
Usually ships in 15 days.
Technical Resources
Related technical articles (Roll over for summary)
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334