Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Thermo Scientific Slide-A-Lyzer Dialysis Kit, 10,000 MWCO, 0.5-3mL, 10 cassettes, 10 buoys, 10 syringes

EW-02905-43
$122.00 USD/EACH

Call 1-888-358-4705 for availability.
Qty

  • High Performance Dialysis require just half the time of dialysis tubing for sample size from 100µl to 30mL
  • Reduces preparation time
  • The innovative cassette design does not require the use of knots or clips that can lead to leaking sample loss.
  • Color coded translucent frame permits smooth sample withdrawal of sub-filter volumes ¨C removing every last drop is easy.
  • High surface area/sample volume ratio will dialyze twice as fast as dialysis via conventional tubing.

  • Available in a variety of sizes
Slide-A-Lyzer® dialysis cassette is ideal for low molecular weight contaminant removal, buffer exchange, desalting and concentration. The cassette membrane is composed of low-binding regenerated cellulose and features a hermetically sealed sample chamber to maintain the highest possible sample retention. Sample introduction and removal are easily accomplished by penetrating the gasket with a hypodermic needle attached to a syringe. The gasket imparts resealability to the needle puncture, ensuring that no sample is lost from the cassette during dialysis.Ready-to-use dialysis devices offer conveniences with preparing, loading and retrieving samples. These devices also increase recovery and decrease sample loss, while increasing sampling efficiency. Choose the ready-to-use device that fits your application. The Micro Float-A-Lyzer is ideal for small sample volumes, below 500µL. The Tube-A-Lyzer uses a flow-through buffer chamber to increase transfer rate and drastically reduce run times. The Float-A-Lyzer G2 makes sample loading and retrieval easy, and includes color coded cap colors for different Molecular Weight Cut-Offs (MWCO). The Thermo Scientific Slide-A-Lyzer has a high surface area to sample volume ratio to increase run times.
Product Type Ready-to-use Dialysis Device
Material Regenerated Cellulose
Membrane MWCO (Daltons) 10000
Min sample volume (mL) 3
Max sample volume (mL) 0.5
Color Orange
Qty/pk 10
Brand Thermo Scientific
Manufacturer number 66382
Model Slide-A-Lyzer Dialysis Cassette Kits

Accessories
Qty/Price
Availability

at $34.00 USD/EACH

Usually ships in 4 days.

at $47.00 USD/EACH

Usually ships in 15 days.

at $27.00 USD/EACH

Usually ships in 15 days.

at $27.00 USD/EACH

Usually ships in 5 days.

at $28.00 USD/EACH

Usually ships in 5 days.

at $29.00 USD/EACH

Usually ships in 15 days.




Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title