United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
3
Representative photo only
Thermo Scientific SnakeSkin Pleated Dialysis Tubing 10 000 MWCO 22 mm dry diameter ID x 35 ft
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-02904-10
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Life Science
>
Dialysis
>
Dialysis Membranes and Tubing
>
Thermo Scientific SnakeSkin Pleated Dialysis Tubes
Share
|
Thermo Scientific SnakeSkin Pleated Dialysis Tubing, 10,000 MWCO, 22 mm dry diameter (ID) x 35 ft
EW-02904-10
$142.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Scale down your dialysis prep time for large sample volumes from 12 to 100 mL
The pleated format makes it easy to open and ready to use, streamlining dialysis preparation, save considerable time over dry, flat tubing.
Regenerated cellulose is ideal for desalting, buffer exchange and removal of small molecular weight contaminants.
The dialysis tubes are supplied in 8¡± sticks containing 35 feet of 22 mm internal diameter tubing. The amount of tubing required can be calculated using 3.7 mL sample per cm of dry tubing. To use it, a researcher simply pulls out the required length of tubing, cuts it off and applies a closure. The sample is then added through the other end of the dry tubing and the second closure is applied.
Specifications
Product Type
Dialysis Tubing
Material
Regenerated Cellulose
Membrane MWCO (Daltons)
10000
Diameter (mm)
22
Length per pack (ft)
35
Brand
Thermo Scientific
Manufacturer number
68100
Model
SnakeSkin Pleated Dialysis Tubes
Accessories
Qty
/
Price
Availability
Dialysis tubing clip, pack of six
EW-02904-15
at
$59.00 USD/PACKAGE
Usually ships in 5 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
High-Performance Dialysis
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title