United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
10 >
Representative photo only
Thermo Scientific Thermolyne Benchtop Muffle Furnace with 350 cu in chamber 120V
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-02053-12
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Heating Equipment
>
Furnaces
>
Muffle Furnaces
>
Thermo Scientific Muffle Furnaces
>
Thermo Scientific Thermolyne Benchtop Muffle Furnaces
Share
|
Thermo Scientific Thermolyne Benchtop Muffle Furnace with 350 cu in chamber; 120V
EW-02053-12
$3,220.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Electronic controls offer programmable operation
Excellent for quantitative analysis and organic and inorganic ashing applications.
Single set point models feature digital temperature control. LED displays chamber temperature continuously; displays set point at the touch of a button. Single set point (SSP) models let you monitor both the set point and actual chamber temperatures. Choose an 8-segment controller (four ramp and four dwell) with additional programming capabilities, and oven temperature protection relay. A safety interlock switch disconnects power to heating elements when door is opened.350 cubic-inch chambers include a ceramic shelf to double your capacity. This model also features a single set point controller with one ramp and one dwell, and an overtemperature protection delay.
What's Included:
a thermocouple sensor and a 6-ft (1.8-m), three-wire power cord with grounded plug.
Specifications
Product Type
Muffle Furnaces
Minimum temperature (º C)
100
Maximum temperature (º C)
1200
Minimum temperature (º F)
212
Maximum temperature (º F)
2192
Temp control accuracy
±3°C
Rise time
80 minutes
Temp uniformity
±3.6°C
Chamber size (" H)
5
Chamber size (" D)
10
Control type
Basic
Manufacturer number
F48015-60
Power (VAC)
120
Power (Hz)
50/60
Model
F48015-60
Brand
Thermo Scientific Thermolyne
Accessories
Qty
/
Price
Availability
Ceramic Hearth Plate
EW-02053-60
at
$56.00 USD/EACH
In stock.
Add to Cart
Ceramic Shelf
EW-02053-61
at
$65.00 USD/EACH
Usually ships in 15 days.
Add to Cart
Technical Resources
Instruction Manual (390 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title