United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
4 >
Representative photo only
Thermo Scientific Thermolyne Compact Benchtop Muffle Furnace 76 cu in 120V
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-33900-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Heating Equipment
>
Furnaces
>
Muffle Furnaces
>
Thermo Scientific Muffle Furnaces
>
Thermo Scientific Thermolyne Compact Benchtop Muffle Furnaces
Share
|
Thermo Scientific Thermolyne Compact Benchtop Muffle Furnace; 76 cu in; 120V
EW-33900-00
$1,585.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Digital control offers simple economical operation
Ideal for ignition tests, gravimetric analysis, and heat treating steel parts
Feature single set point digital control
Digital LED displays chamber temperature
Furnaces feature ceramic fiber insulation for fast heat-up and reduced energy consumption—rise time to 1100degC takes only 50 minutes. Embedded heating elements on three sides give an improved temperature gradient. Drop-down door serves as a convenient shelf for loading and unloading samples. A safety interlock switch disconnects power to heating elements whenever the door is opened. Chambers are constructed of heavy-duty welded steel.
What's Included:
6-ft (1.8-m), three-wire power cord with plug.
Specifications
Product Type
Muffle Furnaces
Minimum temperature (º C)
100
Maximum temperature (º C)
1100
Minimum temperature (º F)
212
Maximum temperature (º F)
2012
Temp control accuracy
±2.0°C
Temp uniformity
±4°C
Chamber size (" H)
3.8
Chamber size (" D)
5
Manufacturer number
FB1315M
Power (VAC)
120
Power (Hz)
50/60
Model
FB1315M
Brand
Thermo Scientific Thermolyne
Accessories
Qty
/
Price
Availability
Ceramic Hearth Plate for item numbers 33900-00, -05
EW-33900-50
at
$24.75 USD/EACH
Usually ships in 11 days.
Add to Cart
Technical Resources
Instruction Manual (260 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title