United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
Representative photo only
Thermo Scientific Thermolyne Extra Large Chamber Muffle Furnace 2765 cu in 208 VAC 50 60 Hz
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-33900-20
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Heating Equipment
>
Furnaces
>
Muffle Furnaces
>
Thermo Scientific Muffle Furnaces
>
Thermo Scientific Thermolyne Extra Large Chamber Muffle Furnace
Share
|
Thermo Scientific Thermolyne Extra Large Chamber Muffle Furnace; 2765 cu in; 208 VAC; 50/60 Hz
EW-33900-20
$11,900.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Triple your already large working area simply by adding shelves
Highly efficient ceramic fiber insulation surrounds the chamber for maximum energy efficiency
Digital LED lets you view actual and set temperatures
This new eight-segment (four ramp, four dwell) programmable muffle furnace offers 2765 cu-in of heated chamber! Simply slide in the two shelves (included) to triple your working area. Heating elements located on sides, top, and bottom of the chamber provide excellent temperature uniformity as well as minimum chamber temperature gradient.
The muffle furnace has safety features that protect your samples as well as the operator. Overtemperature protection cuts off power to the heating elements when the chamber temperature exceeds the set point; door safety switch shuts off the power to the heating elements whenever the door is opened. A 1" diameter vent at the top of the furnace removes contaminants and moisture. The furnace also features a 3/8" diameter port for an external temperature measuring device and an air inlet port for purging.
Specifications
Product Type
Muffle Furnaces
Maximum temperature (º C)
1093
Maximum temperature (º F)
1999
Rise time
151 min at 1000°C
Temp uniformity
±2.0°C at 100°C
Chamber size (" H)
14
Chamber size (" D)
14
Manufacturer number
F30428C-80
Power (VAC)
208
Power (Hz)
50/60
Model
F30428C-80
Brand
Thermo Scientific Thermolyne
Accessories
Qty
/
Price
Availability
Ceramic hearth plate for Large Chamber Muffle Furnace; Measures 5-7/8"W x 3/4"H x 6-3/4"D
EW-33900-54
at
$60.90 USD/EACH
Call 1-888-358-4705 for availability.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title