United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
EW-54102-00
Thickness Gauge, for Paint and Powder Coating
Click to enlarge
More Product Details
Specifications
Calibrations
Reconditioned & Like New
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-54102-00
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-69
NIST-Traceable Calibration; Coating Thickness Gauge (mechanical type)
$99.00 USD/EACH/EACH
Usually ships in 8 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Material Testing & Physical Testing
>
Surface Testing
>
Thickness Gauges
>
Paint and Powder Coating Thickness Gauge
Share
|
Thickness Gauge, for Paint and Powder Coating
EW-54102-00
$759.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Wide range unit provides repeatable precision measurements
Fast and accurate reading rate – greater than 60 readings per minute
Automatically switches to ferrous or non-ferrous for additional flexibility
Intuitive on screen instructions for easy set up and use
This easy to use, low cost gauge provides a simple, accurate, and reliable way to measure
coatings on smooth ferrous (steel) and non-ferrous (aluminum) substrates. The gauge is easy to use and provides on screen instructions guiding you through all of the functions in over 20 user selectable languages. The integral Bigfoot
probe features a v-groove design allowing you to accurately position on smooth, flat surfaces ensuring repeatable measurements. User selectable metric or imperial units display measurements in microns or mils. The ergonomic design makes the gauge comfortable and easy to hold, and the large easy-to-read angled LCD display is perfect for viewing from all angles.
Unit features auto switching to ferrous or non-ferrous capability making it ideal for measuring paint or powder on both steel and aluminum surfaces. The zero cal feature allows for easy calibration and gauge includes a factory calibration reset if you have no access to an uncoated substrate. Constructed form rugged ABS plastic, this gauge is ideal for any paint and powder coating environment.
Can be used in accordance with:
BS 5411 (11) (3), BS 3900 (C5), BS 5599, BS EN ISO 1461, ISO 2178, ISO 2360, ISO 2808 SSPC-PA2 2004, DIN 50984, DIN 50981, ASTM B 244, ASTM B 499 , ASTM D 1400, and ASTM D 1186.
What's Included:
54102-00 includes four check foils (50, 125, 500 & 1000 µm / 2, 5, 20 & 40 mils), soft carry case, and two AAA batteries. 54102-01 includes four check foils (25, 75, 125 & 250 µm / 1, 3, 5 & 10 mils), ferrous checkpiece, non-ferrous checkpiece, soft carry case, and two AAA batteries.
Specifications
Product Type
Thickness Gauge
Description
Paint and powder gauge
Accuracy
±3% or ±3 µm (±0.12 mils)
Operating temperature
32 to 120°F (0 to 50°C)
Dimensions (" W)
3
Dimensions (" H)
4.25
Dimensions (" D)
1.25
Power
two AAA batteries (included)
Display
LCD
Manufacturer number
A415FNFI1
Brand
ELCOMETER INC
Model
415
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Coating Thickness Gauge (mechanical type)
EW-17101-69
at $99.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Reconditioned & Like New
Qty
/
Price
Availability
RECOND 54102-00
EW
-
00299HT
at
$494.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title