United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
Representative photo only
Three Zone Tube Furnace 3800 watts 208 240 VAC
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-33851-25
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Heating Equipment
>
Furnaces
>
Tube Furnaces
>
Thermo Scientific Lindberg Blue M Three-Zone Tube Furnaces
Share
|
Three-Zone Tube Furnace; 3800 watts; 208/240 VAC
EW-33851-25
$7,310.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Temperature uniformity ±1.0°C over 24 inches!
Control each zone independently
Accept multiple tube sizes for versatility
These three-zone tube furnaces feature three 16-segment programmable controllers—one for each zone. Microprocessor-based self-tuning PID control provides excellent control without over-shooting of set temperature. Protect your samples with the adjustable high limit overtemperature protection feature. Furnaces have a uniform temperature throughout the length of the process tube and excellent radial and linear temperature uniformity. The digital LED displays your set and actual temperatures at the same time.
Order process tubes separately.
What's Included:
Type K thermocouple (one per zone) set of tube adapters for the largest size tube the furnace will accept. Order adapters for different sizes of process tubes separately.
Note:
Cord is not included.
Specifications
Product Type
Tube Furnace
Temperature range (º C) (low)
200
Temperature range (º C) (high)
1100
Temp uniformity (º C)
±4°C at 200°C
Chamber size (" L)
6, 12, 6
Chamber size (L (mm))
152. 305, 152
Power (VAC)
208/240
Overall dimensions (" W)
35
Overall dimensions (" H)
21
Overall dimensions (" D)
17
Power (Hz)
50/60
Power (watts)
3800
Shipping weight (lbs)
225
Shipping weight (kg)
102
Brand
Thermo Scientific Lindberg
Model
STF55346C
Manufacturer number
STF55346C
Accessories
Qty
/
Price
Availability
Moldatherm tube adapter for 33851-25, 1"
EW-33851-50
at
$277.00 USD/EACH
Usually ships in 6 days.
Add to Cart
Moldatherm tube adapter for 33851-25, 2"
EW-33851-52
at
$200.00 USD/EACH
Call 1-888-358-4705 for availability.
Add to Cart
Daisy-chain cable to connect multiple units with COM option
EW-33910-52
at
$118.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title