United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
4 >
Representative photo only
Torbeo Hand Held Homogenizer capacity 100 to 1000 mL stator size 2 5 cm 110 VAC 1 2 A
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-04751-10
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Mixers and Blenders
>
Homogenizers
>
Cole-Parmer Homogenizers
>
Cole-Parmer Handheld Homogenizers
Share
|
Torbeo Hand-Held Homogenizer; capacity 100 to 1000 mL; stator size; 2.5 cm; 110 VAC, 1.2 A
EW-04751-10
$926.00 USD/EACH
Usually ships in 6 days.
Qty
:
Add to Cart
Save to shopping list
Mix, emulsify, shred, or chop 1- to 100-mL biological samples
These homogenizers are lightweight and simple to use—just hold homogenizer in a sample vessel and turn on the two-speed switch (7000 or 10,000 rpm). One-handed operation with any suitable beaker or vessel with a 2 inch opening. Material is rapidly dispersed and disintegrated by the ultrahigh-frequency mechanical treatment. No sample transfer loss to transfers from a blender vessel.
What's Included:
a 14.5 cm long probe, stainless steel rotor stator, and a stand. The 110 VAC models include a 5-ft cord with three-prong plug; 230 VAC models include a 5-ft cord only.
Specifications
Product Type
Homogenizer
Max sample size (mL)
1,000
Capacity (mL)
1000
Speed (rpm)
10,000
Brand
Biospec Products
Power (VAC)
110
Power (Hz)
50/60
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
The Use of Steel-Tip Homogenizer Probe Results in Sample Carryover Contamination
Smart Mixing
Mixer Selection Guide
Show All...
Positioning a Mixer in a Cylindrical Vessel
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title