United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
14 >
EW-68900-50
Transducer, 30 PSI, 4-20 mA
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
More info
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-68900-50
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17090-29
NIST-Traceable Calibration; Pressure Transmitter
$155.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pressure & Vacuum Instruments
>
Pressure Transmitters and Pressure Transducers
>
Ashcroft G2 Pressure Transducers
Share
|
Transducer, 30 PSI, 4-20 mA
EW-68900-50
$157.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Affordable performance in a compact, durable package
±1% total error band accuracy
High EMI/RFI rating
High temperature capability
High accuracy is accomplished by marrying a high performance ASIC to a very stable polysilicon thin film pressure sensor. The sensor is electron-beam welded to a pressure fitting of stainless steel, providing high overpressure capability and outstanding durability against shock and vibration. Transducer circuitry is held securely within an internal cage and housed in a reinforced nylon enclosure.
Specifications
Product Type
Pressure transducers
Media compatibility
any media compatible with 17-4PH stainless steel
Wetted materials
17-4 PH SS
Accuracy
±1% of span
Output
4 to 20 mA
Range
0 to 30 psi
Max pressure (psi)
30
Process connection
1/4" NPT(M)
Electrical connections
3-pin Metri-Pak with 3-ft (1-m) cable, 24AWG
Power
9 to 36 VDC
Case
nylon
Brand
Ashcroft
Manufacturer number
G2-7-M02-42-G2-30#-G
Model
G230PSI
Make
TypeG2
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Pressure Transmitter
EW-17090-29
at $155.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Instruction Manual (271 KB)
Virtual Catalog (PDFs)
13-14 Catalog
More info
Dimensional Drawing
Pressure Gauge Selection Made Easy!
Product Manual
Related technical articles (Roll over for summary)
Absolute, Gauge, and Differential Pressure Scales
Common Pressure Terms
Pressure Unit Conversion Constants
Show All...
The Hidden Costs of Mercury-Filled Vacuum Gauges
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title