Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Tri-Flow Lubricant; 12-oz aerosol, case of 6.

EW-08809-15
$89.00 USD/PACKAGE of 6

  • Lubricant with PTFE displaces moisture, prevents corrosion, and will not attract dirt and dust
  • Effective temperature range of -60 to 475°F (-51 to 246°C)
  • Far exceeds the performance of competitive lubes under heavy load, high wear, and extreme temperatures
  • Reduces breakdowns and extends the life of bearings, joints, hinges, cutting tools, cables, gears, and chains
  • Formula is nonstaining and safe on metals, wood, plastics, and rubber

This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Description Tri-Flow® Lubricant
Temp range -60 to 475°F
(-51 to 246°C)
Quantity Case of 6
Packaging 12-oz aerosol
Brand Tri-Flow
Manufacturer number 20006TF
Model 20006TF





Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title