Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Tris Hydrochloride (Small White Flakes/Molecular Biology) (1kg)

EW-88057-53
$206.00 USD/EACH

  • Assay - ≥99%
  • IR - Conforms to standard
  • Melting Point - 152°±2°C
  • Absorbance of a 1M Solution at 280nm - ≤0.05
  • Calcium - ≤10ppm
  • Copper - ≤1ppm
  • Iron - ≤2ppm
  • Magnesium - ≤10ppm
  • Lead - ≤5ppm
  • Arsenic - ≤10ppm
  • Barium - ≤1ppm
  • Manganese - ≤1ppm
  • Electrophoresis - To pass test
  • DNase - Not detected
  • RNase - Not detected
  • Protease - Not detected

This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Chemical Bio-Reagents
Purity grade Molecular Biology
CAS Number 1185-53-1
Form Flakes
Formula weight 157.60
Empirical formula C4H11NO3.HCl
Linear formula H2NC(CH2OH)3•HCl
Synonyms Tris(hydroxymethyl)aminomethane Hydrochloride, Trizma Hydrochloride
Packaging Poly Bottle
Storage symbol Gray
Quantity 1kg





Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title