log in or register
Tub assembly, sealed, 5 L
EW-02658-64
$3,152.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
    Sealed tubs reduce sample loss - ideal for drying smaller particles. To avoid overflow of sample into the filter bag, the filter plate seals on a silicon O-ring, retaining the sample in the tub.What's included: a standard filter top cap. Optional filter top caps are sold separately.
    Description Single, 5 L
    Materials of construction Glass
    Accessories
    Qty/Price
    Availability
    at $786.00 USD/EACH
    Call 1-888-358-4705 for availability.
    at $955.00 USD/EACH
    Call 1-888-358-4705 for availability.
    at $786.00 USD/EACH
    Call 1-888-358-4705 for availability.
    at $786.00 USD/EACH
    Call 1-888-358-4705 for availability.
    Build a complete system
    Technical Resources
    Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Leasing Available
    Financing is avaliable for many of our products.
    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title