United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
6 >
SI-06413-40
Tygon® R-3603 Vacuum Service Tubing
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
SI-06413-40
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Tubing, and Hose
>
Tubing
>
Flexible Tubing
>
Tygon Tubing
>
Tygon Vacuum Tubing
Share
|
Sorry, this item is now discontinued but has a suggested replacement.
You can compare specifications to the suggested replacement SI-0641440
,
browse similar items
or
Contact us for more information.
Tygon vacuum tubing, 1/2"ID x 1-1/8"OD, 10 ft/pack
SI-06413-40
$141.00 USD/PACKAGE
Quantity Discounts
Order
Discount
Price
[X]
4
10%
$126.90/PACKAGE
10
15%
$119.85/PACKAGE
20
20%
$112.80/PACKAGE
Discontinued
Qty
:
Save to shopping list
Formulation:
TYGON R-3603 vacuum
Applications:
Vacuum service—withstands 29.9" Hg at 75°F (24degC)
Characteristics:
Heavy wall, nontoxic, and non-oxidizing. Transparent.
Certification:
Tubing is manufactured according to GMP.
Temperature range:
–58 to 165°F (–50 to 74°C)
Sterilize:
With ethylene oxide or autoclave for 30 min at 250°F, 15 psi.
Specifications
Product Type
Tubing
Working pressure (psi)
45
Maximum temperature (º F)
165
Minimum temperature (º F)
-58
Tubing ID (in.)
0.5
Tubing ID (mm)
12.8
Tubing OD (in.)
1.125
Tubing OD (mm)
28.6
Max psi
45
Certifications
GMP
Translucent
Yes
Sterilizable
Yes
Color
Transparent
Feet per pack
10
Vacuum rating
29.9" Hg
Tubing formulation
Tygon
Brand
Tygon
Manufacturer number
AACUN040-CP
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Cole-Parmers Bioconnect Program
Decrease bacterial growth in your tubin
The STAMP Tubing Selection Guide
Show All...
Tygon® Flexible Plastic Tubing to Tubing Solvent Sealing
Tubing Selection Guide
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title