Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Tyvek cleanroom frock; XL, 30/case

EW-33683-06
$240.00 USD/CASE

  • Superior particulate and light barrier protection
Tyvek® Apparel
TYVEK apparel is processed in a certified Class 10 cleanroom with ultraclean water to meet USP standards for purified water and microelectronics grade P-1 water as proposed by ASTM. These cleanroom garments are double packaged to meet Class A standards. They are first bagged and sealed individually, then packed in a case with a polyplastic liner. Frocks feature serged seams; all other TYVEK apparel features bound seams. A serged seam is a basic stitched seam that is appropriate for exposure to nonhazardous substances. A bound seam is reinforced with binding for strength and tear resistance.IsoClean® Tyvek® Frocks have five snap closures, elastic wrists, and a three-snap adjustable mandarin collar. Serged seams.
Product Type Lab Coats
Description Elastic wrists & adjustable collar
Size X-Large
Color White
seam Serged
Material Polyethylene
Compliance ISO Class 4 (formerly Class 10)
Quantity 30/cs
Manufacturer number IC270BWHXL003000
Brand Tyvek
Model IC270BWH003000





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title