dummy
EW-97651-10
- With interchangeable NIST-traceable sensors
This radiometer is used to monitor the useful life of any UV light source by measuring lamp irradiance. It operates with a 9 V battery for up to 120 hours. Select three intensity ranges: 0 to 20 mW/cm2, 0 to 2000 µW/cm2, and 0 to 200 µW/cm2. Optional 10:1 attenuator extends the reading range up to 200 mW/cm2.
Choose one of the three interchangeable sensors for measuring 254 nm, 302 nm, or 365 nm ultraviolet wavelengths (sold separately). All sensors are calibrated with NIST traceability, and are ±5% accurate. The 3-ft cable length is ideal for measuring UV in remote locations.
| Product Type |
Ultraviolet Meters/Detectors |
| Display |
3.5 digit |
| Dimensions |
6.5 in L x 3.375 in W x 2 in H |
| Power |
9 V battery (sold separately) |
| Manufacturer number |
97-0015-02(UVX) |
| Model |
97-0015-02(UVX) |
at $25.60 USD/PACKAGE of 4
In stock.
at $322.00 USD/EACH
In stock.
at $322.00 USD/EACH
Usually ships in 13 days.
at $54.00 USD/EACH
Usually ships in 10 days.
Technical Resources
Related technical articles (Roll over for summary)
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334