dummy
EW-50513-02
Usually ships in 8 days.
Qty:
- Measure damaging UV (ultraviolet) light
Conduct UV degradation studies for pharmaceutical, cosmetic, semiconductor, printing, forensic applications and more. The UV light intensity appears on the large backlit display in either ìW or mW/cm² while the small lower display simultaneously shows minimum, maximum, average, and recorded data. Up to 20 data points can be recorded automatically at a variety of intervals. The
UV-A/B model measures long and medium wave UV-A/B for forensics, chromatography, electrophoresis, and dermatology applications. Use the UV-C model for UV light fume hoods, UV sterilization, industrial glass, and nondestructive testing.
What's Included:
One UV probe, tripod screw, magnetic mount, hard foam-lined carrying case, and one 9 V battery.
| Product Type |
Ultraviolet Meters/Detectors |
| Description |
Light meter, UV-A/B |
| Display |
dual-line backlit LCD |
| Function |
min/max/avg, zero point reset, hold, low battery, data storage/recall |
| Output |
none |
| Accuracy |
±4% ±1 digit (at 23±5,°C) |
| Range |
1 µW/cm2 to 40.00 mW/cm2 |
| Resolution |
1 µW and 0.01 mW/cm2 |
| Dimensions |
2 in W x 6 in H x 1.5 in D (5 x 15.2 x 3.8 cm) |
| CE Compliance |
Yes |
| Power |
One 9 V battery |
| Manufacturer number |
850010 |
| Model |
850010 |
at $25.60 USD/PACKAGE of 4
In stock.
Technical Resources
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334