United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths & Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cleaning & Janitorial Supplies
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Lab Supplies & Consumables
Laboratory Equipment
Life Science
Masterflex® Peristaltic Pumps
Masterflex® Pump Tubing
Material Storage & Handling
Material Testing & Physical Testing
Microscopes & Magnification
Mixers and Blenders
pH, Conductivity, Dissolved Oxygen, and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity and Moisture Products
Tools
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 15
of
16 >
EW-99280-57
Ultraclear Tap Water Feed System, Uv/uf/tm, 60l Tank
Representative photo only
Click to enlarge
More Product Details
Specifications
Build a complete system
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-99280-57
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Laboratory Equipment
>
Water Purification Systems and Accessories
>
Water Deionization Systems
>
SG Water Ultra Clear TWF
Share
|
SG Water Ultra Clear TWF
-
Ultraclear Tap Water Feed System, Uv/uf/tm, 60l Tank
EW-99280-57
$10,000.00 USD/EACH
Usually ships in 53 days.
Qty
:
Add to Cart
Save to shopping list
Obtain ultrapure, deionized water from a tap water feed
œ Exceed ASTM, NCCLS, and ISO Type 1 requirements
œ Flexible-hose dispenser lets you remotely dispense water from up to two feet away
œ Models with TOC monitor let you continuously monitor TOC levels to <1 ppb,
resistivity, conductivity, and temperature on the easy-to-read display
These water purification systems are designed to take municipal tap water and produce reagent grade
ultrapure water without the need for multiple systems—reducing operating costs. The
pretreatment, RO module, deionization cartridge, storage tank, UV oxidation chamber, polisher,
ultrafilter, and sterile filter are all integrated into one convenient unit. Dispense rate is 1.8 L/min.
Automatic flushing mechanism prevents contaminant build-up and extends service life. Unit can
be either bench or wall mounted; 100 to 240 VAC, 50/60 Hz operation. Cartridge changes are quick
and simple.
Specifications
Capacity
60L
Max flow rate
1.8 L/MIN
Dimensions
21" X 36" X 13"
Resistivity threshold
18.2
Description
Tap Water Feed, UV/UF/TM Module
Manufacturer number
2004-TM-D/60
Model
2004-TM-D/60
Technical Resources
Virtual Catalog (PDFs)
09-10 Catalog
11-12 Catalog
Related technical articles (Roll over for summary)
Water Purification
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Basics of Cartridge Filtration
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Isolating Genomic DNA from Whole Blood
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Job Title
|
Category
|
Brand
|
Part Number
|
Price
View product categories by:
Popularity
|
Favorites
|
Job Title
|
Price
|
Part Number
View popular items by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Job Title
View reviews by:
Popularity
|
Product Title
|
Best Rated