Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Uvex Genesis Black frame glasses with clear lenses

EW-86325-70
$8.75 USD/EACH

quantity discounts Quantity Discounts
Order Discount Price
[X]
10 10% $7.88/EACH

  • Lenses exceed, by seven times, the standards set by ANSI for impact velocity
  • Sleek, stylish design for cool, comfortable protection
Wraparound lens design provides uncompromised peripheral vision. Internal points of contact serve as a shock absorber to diffuse impact energy away from eyes.
A ventilation channel between the frame and lens allows moisture to escape— preventing fogging. Coating helps to keep your lenses scratch free. Lenses and temple stems are fully adjustable to provide a custom fit.
Compliance: ANSI Z87+ (High Impact), CSA Z94.3, military V0
Product Type Glasses
Color Black
Lens Clear
Compliance ANSI Z87+ (High Impact), CSA Z94.3, military V0
Brand Uvex
Model S3200
Manufacturer number S3200





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title