- Detects invisible food residue without an instrument -in only 10 to 60 seconds
VERIcleen is sensitive to food residues including meat, dairy, bakery, beverages, eggs, fish, etc. It is an inexpensive rapid hygiene program to the food service, retail and institutional industries. Applications include HACCP programs, hand washing hygiene compliance, and overall sanitation monitoring. Application is extremely simple, just spray to wet dry surface and swab. A ''clean surface'' shows no purple until after 3 minutes. A ''Not clean'' surface shows purple in 10 to 60 seconds.
Each kit includes 50 tests in a light-tight snap-top container, 1 bottle of wetting liquid with atomizer, instruction leaflet, and desiccant packet (if desiccant inside the packet has turned from blue to pink, the strips have been stored improperly, and may be invalid).