United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
4 >
Representative photo only
Vacuum Oven 2 3CU FT 120VAC
Click to enlarge
More Product Details
Specifications
Calibrations
Reconditioned & Like New
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-52402-96
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17002-30
Innocal NIST Calibration for Ovens
$160.00 USD/EACH/EACH
Usually ships in 17 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Heating Equipment
>
Ovens
>
Vacuum Ovens
>
Lab Companion Vacuum Ovens
Share
|
Vacuum Oven 2.3CU.FT; 120VAC
EW-52402-96
$4,050.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Feature wide temperature range for a variety of test applications
Perform a variety of applications from moisture determination, aging test, and sterilization to solvent separation under controlled atmosphere conditions
Ideal for separating solvents from a solution without high-temperature drying
Heat-tempered full-view window allows continuous monitoring of chamber
User-friendly digital control panel simplifies operation
Separate vacuum and vent ports allow greater control of vacuum rates. Vacuum levels are maintained by a tightly sealed door gasket and push-button latch. Block heaters and anodized aluminum plate shelves provide uniform heating throughout the oven chamber.
What's Included:
shelves and 120 VAC or 230 VAC power cord.
Specifications
Product Type
Mechanical Convection Oven
Capacity (Liters)
56.62
Capacity (cu ft)
2
Chamber size (" W)
15.75
Resolution
1°C
Chamber size (" H)
15.75
Chamber size (" D)
16
Minimum temperature (º C)
Ambient ±5
Maximum temperature (º C)
250
Minimum temperature (º F)
41
Maximum temperature (º F)
482
Power (VAC)
120
Temp uniformity
±1.5°C at 150°C
Temp control accuracy
±2.0°C
Rise time
100°C within 85 min
Shpg wt (lbs)
227 (103 kg)
Vacuum display
Analog
Display
Digital LED
Vacuum range
0 - 0.1 MPa
Control type
PID microprocessor
Power (Hz)
50/60
Power (Amps)
11.7
No. of shelf positions
3
No. of shelves
3
Brand
Lab Companion
Construction
Stainless steel interior
Overall dimensions
31-3/4"W x 23-1/2"H x 22"D
(80 x 60 x 56 cm)
Model
AAH13126U
Manufacturer number
AAH13126U
Calibrations
Qty
/
Price
Availability
Innocal NIST Calibration for Ovens
EW-17002-30
at $160.00 USD/EACH
Usually ships in 17 days.
Add to Cart
Reconditioned & Like New
Qty
/
Price
Availability
RECOND 52402-96
EW
-
00274WH
at
$2,335.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Taking the “Dry” Out of Moisture Determination
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title