United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
9 >
EW-07061-11
Image is for illustrative purposes only. Please refer to product description
Click to enlarge
More Product Details
Specifications
Technical Resources
Flow Charts
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-07061-11
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Vacuum Pumps
>
Dry Vacuum Pressure Pumps
>
High-Capacity Vacuum/Pressure Pumps
Share
|
Vacuum/pressure diaphragm pump with vacuum gauge, dual/series head, 1.15 cfm, 115 VAC
EW-07061-11
$650.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Free-air capacities up to 2.2 cfm (62.3 L/min)
Shaded-pole fan-cooled motor has thermal overload switch to protect your pump system.
Heads on pumps 07061-00 and -02 are connected in parallel for higher capacity; heads on 07061-10 and -12 are connected in series for greater vacuum.
Select pumps with or without gauges, regulators, and valves.
What's Included:
6-ft, three-wire cord; the 115 VAC models include U.S. standard plug.
Specifications
Max vacuum
29.0" Hg
Product Type
Vacuum/Pressure Diaphragm Pump
Free-air capacity
1.15 cfm (32.5 L/min)
Port size
1/4" NPT(F), 3/8" hose barb on gauges
Max temperature
100°F (38°C)
Motor type
permanent split capacitor
Duty cycle
continuous
hp
1/4
Wetted parts
aluminum stainless steel, NORDEL®, and NOMEX®
Dimensions
11-13/16"L x 5-1/2"W x 10-1/4"H
Amps
3.6
Power
115 VAC, 60 Hz
Stages
Dual/series
Technical Resources
CP_Flow_Curve_07061-11.pdf
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Basics of Cartridge Filtration
A Vacuum Pump That Survives Freeze Dryer Applications
Diaphragm Vacuum Pump Buyer's Guide
Show All...
Vacuum/Compressor Pump Selection Guide
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title