United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
6 >
EW-79200-30
Vacuum/Pressure Pumps Coated with PTFE
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-79200-30
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Vacuum Pumps
>
Dry Vacuum Pressure Pumps
>
Vacuum/Pressure Pumps Coated with PTFE
Share
|
Vacuum/pressure diaphragm pumps, PTFE-coated wetted parts, 0.75 cfm, 115 VAC
EW-79200-30
$340.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Wetted parts are coated with PTFE to handle your toughest applications
Wetted parts are coated with PTFE to handle your toughest applications.
Lightweight aluminum construction and compact size make pumps ideal for benchtop applications such as vacuum filtration.
Pumps with gauges and regulators give you more control of your vacuum/pressure applications.
Pumps include suction cup feet and a 6-ft power cord with built-in switch (the 115 VAC models have grounded plug, 220 VAC and DC models have stripped ends).
Specifications
Max vacuum
23.2" Hg
Free-air capacity
0.75 cfm (21.2 L/min)
Port size
1/8" NPT(F)
Max temperature
104°F (40°C)
Motor type
shaded pole
Duty cycle
continuous
hp
1/20
Wetted parts
PTFE FTFE
Dimensions
6-7/8"L x 4-1/4"W x 4-3/4"H
Amps
1.7
Power
115 VAC, 60 Hz
Accessories
Qty
/
Price
Availability
Vacuum Gauge with Digital Dial and Reading
EW-07379-20
at
$935.00 USD/EACH
In stock.
Add to Cart
Vacuum Meter/Controller with Digital Dial and Reading, 115 VAC
EW-07379-40
at
$2,320.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Basics of Cartridge Filtration
A Vacuum Pump That Survives Freeze Dryer Applications
Diaphragm Vacuum Pump Buyer's Guide
Show All...
Vacuum/Compressor Pump Selection Guide
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title