Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Sorry, this item is now discontinued. Please browse similar items.
or Contact us for more information.
Variable-Speed Microcentrifuge, Purple, 230V
Item# YO-17309-35
$1,757.00 USD/EACH
Discontinued
Qty
  • Brushless motor is maintenance free; economically priced!
  • Wide speed range meets a variety of applications including microfiltration and protein or nucleic acid separations.
  • Maintenance-free brushless motor accelerates to maximum speed in only 10 seconds; decelerates in 9 seconds.
  • Unique airflow pattern and ventilated chamber keeps sample temperature stable throughout run.
  • Durable, analog controls are positioned up front for ease of use.
  • Use for momentary, continuous, or timed runs.
Centrifuge comes with 18 x 1.5/2.0-ml fixed-angle rotor.
What's Included:
All models include a 6-ft cord; 120 VAC models include a three-prong plug; 230 VAC models include a European plug
Product Type Microcentrifuge
Max rpm 14,000
Max RCF 16,000 x g
Rotor Fixed Angle
Color Purple
Speed range 0 to 14,000 rpm
Max capacity 25mL
Refrigerated No
Tube capacity 18 x 1.5/2.0-mL
Power (VAC) 230
Rotor (included) Yes
Timer 1 to 30 minutes
Capacity 1.5-2.0mL
Power (Hz) 50/60
Max RCF (x g) 16,000
Dimensions 8 1/4"W x 7"H x 8 15/16"D
Calibrations
Qty/Price
Availability
at $120.00 USD/EACH
Usually ships in 8 days.
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews