log in or register
Vortex-Genie 2 Shaker, 120 VAC
EW-04724-00
$355.00 USD/EACH
Usually ships in 9 days.
Qty
  • Enhanced vortex action for maximum high-speed touch mixing
  • Solid metal casing with rubber feet prevents shaker from ''walking'' across your benchtop
Vortex-Genie® 2 Shaker allows you to control speed intensity by varying pressure to the head. The variable speed shaking allows you to adjust the level of mixing from gentle shaking to vigorous vortexing. Easy-to-change platforms allow you to adapt your shaker to a variety of applications. Use the pop-off cup for single tube shaking or the 3'' platform for multiple tube or flask shaking.
What's Included:
Pop-off cup and 3" platform. Power cord and plug (Euro plug on 230 VAC model).
Product Type Vortex Shakers
Min speed (rpm) 600
Max speed (rpm) 3200
Platform dimensions 1-1/8" dia pop-off cup, 3" dia platform
Dimensions 4-7/8"W x 6-½"H x 6-½" D
Brand Scientific Industries
Power (VAC) 120
Power (Hz) 60 Hz
Power (Amps) 0.65 A
Accessories
Qty/Price
Availability
Platform Starter Kit
EW-04724-50
at $80.00 USD/EACH
In stock.
at $45.50 USD/PACKAGE of 2
In stock.
at $50.00 USD/EACH
Usually ships in 15 days.
at $97.00 USD/EACH
Call 1-888-358-4705 for availability.
at $142.00 USD/EACH
Usually ships in 8 days.
at $225.00 USD/EACH
Usually ships in 4 days.
at $35.00 USD/EACH
In stock.
at $75.00 USD/EACH
Usually ships in 6 days.
at $85.00 USD/EACH
Usually ships in 6 days.
at $95.00 USD/EACH
Usually ships in 6 days.
at $39.00 USD/EACH
Usually ships in 6 days.
at $33.00 USD/EACH
In stock.
PLATFORM STARTER KIT
EW-04724-80
at $142.00 USD/EACH
Usually ships in 10 days.
at $41.00 USD/SET of 1
Usually ships in 8 days.
at $7.00 USD/SET of 1
Usually ships in 6 days.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title