Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
W A Hammond Drierite Desiccant Bags, 2 oz/bag, 400 bags/pk
W A HAMMOND DRIERITE CO  |   Mfr# 60012  |   Item# EW-07193-16
$208.00 USD/PACKAGE
Are you a reseller or buying in bulk? Call us today.
Usually ships in 10 days.
Qty
  • Desiccant bag will reduce the humidity to a 100°F dew point in 10 hours
  • These bags are dust-tight and will protect for many months
  • Materials packaged with these bags are protected from corrosion
These nonwoven crepe bags are filled with Drierite (anyhydrous calcium sulfate) to completely dry the air within a package or container during shipment or storage. The bags are sturdy and held together by heat seals. Drierite bags also perform well in elevated temperatures and will not release absorbed moisture at less than 350°F.
Product Type Desiccant
Qty/pk 400
Model 60012
Manufacturer number 60012
Brand W A HAMMOND DRIERITE CO
Dimensions (" L) 4.75
Dimensions (" W) 3.75
Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456