United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
Representative photo only
W S Tyler Ro Tap E Sieve Shaker for 8 diameter sieves 110 VAC 60 Hz
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-59996-68
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Sieves
>
Sieve Shakers
>
Ro-tap, Ro-tap II, and Ro-tap E Sieve Shakers
>
W.S. Tyler Ro-Tap E Sieve Shakers
Share
|
W.S. Tyler Ro-Tap E Sieve Shaker, for 8" diameter sieves, 110 VAC, 60 Hz
EW-59996-68
$4,410.00 USD/EACH
Usually ships in 14 days.
Qty
:
Add to Cart
Save to shopping list
Reproduce the rotating and tapping motion of hand-sieving but with more uniform motion for highly reproducible results
Ro-Tap
®
E Sieve Shakers for 8'' dia sieves hold up to six full-height or 13 half-height sieves with top cover and receiving pan
Shakers are actuated by a built-in, 99-minute digital timer with 0.1 second accuracy. They are powered by a 1/4-hp motor and include a 5-ft cord (110 VAC models also include U.S. three-prong plug).
Ro-Tap
®
E Sieve Shakers for 8'' sieves are stand-alone benchtop units that are virtually maintenance-free, offer quiet operation and can be operated directly on a laboratory countertop. Options include automatic controlled amplitude, and fine and coarse analysis settings.
Specifications
Product Type
Sieve Shakers
Sieves size accepted (" OD)
8"
rpm
1750
Dimensions (" W)
18
Dimensions (" H)
18
Dimensions (" D)
30
Dimensions (cm)
45.7 W x 45.7 W x 76.2
Power (Hz)
60 Hz
Timer range (min)
99
Power (VAC)
110
CE Compliance
Yes
Shipping weight (lbs)
80
Shipping weight (kg)
36
Brand
W.S. Tyler
Manufacturer number
RX-29-E
Accessories
Qty
/
Price
Availability
W.S. Tyler Sound Enclosure for Ro-tap Sieve Shaker, 34"W x 30-1/2"H x 24"D
EW-59986-65
at
$810.00 USD/EACH
In stock.
Add to Cart
W.S. Tyler Test Stand for Ro-Tap Shakers, 20
EW-59986-98
at
$356.00 USD/EACH
In stock.
Add to Cart
W.S. Tyler Wet Test Kit for 8" Diameter Ro-tap Shakers Only
EW-59986-99
at
$846.00 USD/EACH
Usually ships in 14 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Nominal Dimensions of Standard Sieves
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title