United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 5
of
12 >
EW-10122-10
Bead bath with beads
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Technical information
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-10122-10
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Baths and Circulators
>
Specialty Baths
>
Waterless Bead/Sand Baths
>
Waterless Bead Baths
Share
|
Waterless bead bath 14L, with Beads 12L
EW-10122-10
$1,899.00 USD/EACH
Usually ships in 4 days.
Qty
:
Add to Cart
Save to shopping list
Low maintenance, eco-friendly alternative to water baths
Eco-friendlybeads are recyclable, use 50% less electricity, and do not require germicides
Beads hold things in place, eliminating the need for racks and weights
Bath is always ready, eliminating warm-up times
Save time--no moreFast heat-ups and accurate temperature control to 600° C
Safer operation than oil baths
Dry, inert, nontoxic aluminum oxide ensures hazard-free operation emptying, cleaning, and refilling
These baths use metallic beads instead of water, delivering exceptional temperature uniformity of +/-1 degree without lengthy warm-up times. Beads create a dry bath, avoiding the contamination and maintenance of traditional water baths. They also improve energy efficiency over 50% of traditional water baths.
Beads hold a variety of containers securely in place without any float-away or ruined samples. You are no longer limited to water-tight containers, so you can incubate multi-well plates, petri dishes, and open-top samples at any angle.
Metallic beads have high thermal conductivity in the range of +/-180° C. Smooth, rounded surfaces measure 5 to 8 mm diameter and 1 to 2 mm high. Beads are moisture and gas impermeable, and are easy to keep clean without harmful germicides. These versatile beads can also be used in a container for freezers, ovens, and incubators.
Includes beads for bath.
What's Included:
stainless steel lid and power cord. Purchase bath
with beads or bath only and beads separately.
Specifications
Product Type
Bead Bath
Capacity (Liters)
14L
Dimensions (" W)
15 7/8" (40.4 cm)
Dimensions (" H)
18 1/2" (47 cm)
Dimensions (" D)
8 1/2" (21.6 cm)
Temp setting
ambient to 80°C
Immersion depth
6" (15.2 cm)
Wetted materials
Stainless steel
Power (VAC)
100 to 120
Power (Hz)
50/60
Power (Amps)
4.5
Uniformity
±1.0°C @ 37°C
Display
LED
Bath opening (" W)
12 1/8" (30.7 cm)
Bath opening (" L)
12" (30.5 cM)
Accessories
Qty
/
Price
Availability
Beads for dry bath, 2-liters
EW-06274-50
at
$195.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Beads for dry bath, 4-liters
EW-06274-51
at
$320.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Beads for dry bath, 8-liters
EW-06274-52
at
$560.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Technical Resources
Solving_Common_Water_Bath_Problems (17 KB)
Bead Bath Manual
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Chiller/Recirculator Selection Guide
Learn how to choose the chiller that best suits all of your cooling needs
Bath and Circulator Specifications Terminology
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title