United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, Fittings, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-90205-22
Waterproof Remote Probe Thermometer
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-90205-22
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17003-00
NIST-Traceable Calibration; Bimetals or Pocket Digital Thermometers
$60.00 USD/EACH/EACH
Usually ships in 8 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Temperature, Humidity, Moisture and Weather Products
>
Temperature Products
>
Digital Thermometers
>
Digital Thermometers - General Purpose
>
Cole-Parmer Mini Digital Thermometers with Remote Probe
>
Cole-Parmer Waterproof Remote Probe Thermometer
Share
|
Waterproof Remote Probe Thermometer
EW-90205-22
$42.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Ideal for measuring temperature from a distance.
Typical response time is 30 seconds for 99% of new reading
Thermometers have a 304 stainless steel penetration stem
All models have a NIST traceable certificate
Battery included
Waterproof Remote Probe Thermometer features a 10-ft flexible cord, degF/degC switchable ranges, a display that updates every second, and Min/Max memory. Perfect for wet areas— case is water/shockproof and probe is impervious to acid and solvents. Supplied with suction cups, magnet, and Velcro
®
tabs to attach to any surface. Use ON/OFF button to conserve battery life.Waterproof Remote Probe Thermometer features a 10-ft flexible cord for measuring temperatures from a distance. Perfect for wet areas, case is water/shock proof, and probe is impervious to acid and solvents. Supplied with suction cups, magnet and VELCRO
to attach to any surface. Model also includes °F/°C switchable ranges, display that updates every second, and min./max. memory. Use ON/OFF button to conserve battery life.
Specifications
Product Type
Digital Indicator
Min temperature (º F)
-58
Min temperature (º C)
-50
Max temperature (º C)
300
Max temperature (º F)
572
Accuracy (º F)
±1.8°
Accuracy (º C)
±1.0°
Display
3.5 digit, 0.25 in
Probe (Remote)
Yes
Probe dimensions
5 in L
Resolution
0.1° F/C
Power
Battery (included)
Model
4039CP
Manufacturer number
4039CP
Accessories
Qty
/
Price
Availability
Batteries, 1.5V button cells, 6/PK
EW-09377-16
at
$15.50 USD/PACKAGE of 6
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Bimetals or Pocket Digital Thermometers
EW-17003-00
at $60.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
11-12 Catalog
13-14 Catalog
Related technical articles (Roll over for summary)
Temperature FAQs
Temperature Instrument Ranges
Getting a Handle on Temperature
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title